Methyl 3-(2-aminophenyl)acrylate 97%

Supplier: Ambeed
Warning

A140635-5G A140635-10G
76871-184EA 141 USD
76871-184 77885-183
Methyl 3-(2-aminophenyl)acrylate 97%
Methyl 3-(2-aminophenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00031415
CAS Number: 1664-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR