5-Bromo-1H-1,2,3-triazole 95%

Supplier: Ambeed
Warning

A140171-100MG A140171-1G A140171-250MG A140171-5G
76871-178EA 14 USD
76871-178 76871-180 76871-182 77549-706
5-Bromo-1H-1,2,3-triazole 95%
5-Bromo-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₂H₂BrN₃
MDL Number: MFCD10696349
CAS Number: 40964-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR