N-(4-(2-Aminoacetyl)-5-methoxy-2-phenoxyphenyl)methanesulfonamide hydrochloride 95%

Supplier: Ambeed
Warning

A139873-100MG A139873-1G
76871-172EA 8 USD
76871-172 76871-174 76871-176
N-(4-(2-Aminoacetyl)-5-methoxy-2-phenoxyphenyl)methanesulfonamide hydrochloride 95%
N-(4-(2-Aminoacetyl)-5-methoxy-2-phenoxyphenyl)methanesulfonamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₉ClN₂O₅S
MDL Number: MFCD12911398
CAS Number: 149436-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR