1-(Pyrimidin-2-yl)piperidin-4-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A139709-100MG A139709-1G A139709-250MG A139709-5G
76871-160EA 29 USD
76871-160 76871-162 76871-164 76871-166
1-(Pyrimidin-2-yl)piperidin-4-amine dihydrochloride 97%
1-(Pyrimidin-2-yl)piperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆Cl₂N₄
MDL Number: MFCD11052947
CAS Number: 1179369-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR