4-(Carboxymethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Boronophenyl)acetic acid

A139637-100MG A139637-1G A139637-250MG A139637-5G
76871-152EA 9 USD
76871-152 76871-154 76871-156 76871-158
4-(Carboxymethyl)phenylboronic acid 98%
4-(Carboxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MDL Number: MFCD03425919
CAS Number: 90111-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR