2-Ethoxy-6-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A139487-1G A139487-250MG A139487-5G
76871-146EA 19 USD
76871-146 76871-148 76871-150
2-Ethoxy-6-(trifluoromethyl)pyridine 98%
2-Ethoxy-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MDL Number: MFCD17015840
CAS Number: 1245563-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR