5-Bromo-1H-pyrrolo[2,3-b]pyridin-6-amine 95%

Supplier: Ambeed
Warning

A139423-1G A139423-250MG
76871-140EA 640.79 USD
76871-140 76871-142
5-Bromo-1H-pyrrolo[2,3-b]pyridin-6-amine 95%
5-Bromo-1H-pyrrolo[2,3-b]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD12962862
CAS Number: 1190322-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR