O-Butylhydroxylamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A139319-10G
A139319-1G
A139319-250MG
A139319-5G
76871-122EA
258.86
USD
76871-122
76871-124
76871-126
76871-128
O-Butylhydroxylamine hydrochloride 97%
O-Butylhydroxylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₁NO·HCl MW: 125.60 g/mol |
MDL Number:
MFCD00487631 CAS Number: 4490-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...