Methyl 1-methyl-1H-indole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Methoxycarbonyl)-1-methyl-1H-indole

A139038-100MG A139038-1G A139038-250MG
76871-108EA 12.74 USD
76871-108 76871-110 76871-112
Methyl 1-methyl-1H-indole-4-carboxylate 95%
Methyl 1-methyl-1H-indole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
MDL Number: MFCD09839048
CAS Number: 1444-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR