2-(4-Bromo-2,5-difluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A138915-250MG A138915-10G
76871-106EA 36.08 USD
76871-106 77611-258
2-(4-Bromo-2,5-difluorophenyl)acetic acid 98%
2-(4-Bromo-2,5-difluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
MW: 251.025 g/mol
MDL Number: MFCD11847567
CAS Number: 871035-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR