N-(5-Methoxy-2-phenoxyphenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A138910-100MG A138910-250MG
76871-102EA 20.16 USD
76871-102 76871-104
N-(5-Methoxy-2-phenoxyphenyl)methanesulfonamide 98%
N-(5-Methoxy-2-phenoxyphenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄S
MW: 293.34 g/mol
MDL Number: MFCD13152085
CAS Number: 123664-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR