1,4-Dihydroxy-9,10-anthraquinone 95%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dihydroxyanthracene-9,10-dione, Quinizarin, 1,4-Dihydroxyanthraquinone

A138561-100G A138561-25G A138561-500G
76871-080EA 16.98 USD
76871-080 76871-082 76871-084
1,4-Dihydroxy-9,10-anthraquinone 95%
1,4-Dihydroxy-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₈O₄
MW: 240.22 g/mol
MDL Number: MFCD00001209
CAS Number: 81-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR