Isopropyl ((S)-((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)-L-alaninate 98%

Supplier: Ambeed
Warning

Synonyms: Tenofovir alafenamide

A138397-100MG A138397-10G A138397-10MG A138397-1G A138397-1MG A138397-250MG A138397-5G A138397-5MG
76871-064EA 71 USD
76871-064 76871-066 76871-068 76871-070 76871-072 76871-074 76871-076 76871-078
Isopropyl ((S)-((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)-L-alaninate 98%
Isopropyl ((S)-((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)-L-alaninate

Precaución: for Research Use Only

Formula: C₂₁H₂₉N₆O₅P
MW: 476.46 g/mol
MDL Number: MFCD23843796
CAS Number: 379270-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR