N-(5-Methyl-1H-pyrazol-3-yl)acetamide 98%

Supplier: Ambeed
Warning

A138323-100MG A138323-250MG
76871-060EA 49.87 USD
76871-060 76871-062
N-(5-Methyl-1H-pyrazol-3-yl)acetamide 98%
N-(5-Methyl-1H-pyrazol-3-yl)acetamide

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MW: 139.16 g/mol
MDL Number: MFCD00114577
CAS Number: 83725-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR