4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

A138141-100MG A138141-1G A138141-250MG A138141-5G
76871-048EA 39 USD
76871-048 76871-050 76871-052 77885-170
4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester 95%
4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₃
MDL Number: MFCD04440855
CAS Number: 364794-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR