5-(Trifluoromethyl)-2-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (5-(Trifluoromethyl)pyridin-2-yl)boronic acid

A138006-250MG A138006-1G
76871-030EA 133.68 USD
76871-030 77566-772
5-(Trifluoromethyl)-2-pyridineboronic acid 95%
5-(Trifluoromethyl)-2-pyridineboronic acid

Precaución: For research use only.

Formula: C₆H₅BF₃NO₂
MW: 190.9156 g/mol
MDL Number: MFCD06227445
CAS Number: 1162257-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR