2,3-Difluoro-4-methylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2,3-Difluoro-4-methylphenyl)boronic acid

A136551-10G A136551-1G A136551-250MG
76870-986EA 105 USD
76870-986 76870-988 76870-990
2,3-Difluoro-4-methylphenylboronic acid 95%
2,3-Difluoro-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₂
MDL Number: MFCD09751348
CAS Number: 508235-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR