4-Bromocarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-9H-carbazole

A136315-10G A136315-5G A136315-1G A136315-25G A136315-250MG
76870-980EA 5.31 USD
76870-980 76870-982 76870-984 77592-570 77592-572 77670-952
4-Bromocarbazole 97%
4-Bromocarbazole

Precaución: for Research Use Only

Formula: C₁₂H₈BrN
MDL Number: MFCD18450175
CAS Number: 3652-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR