5-Bromobenzo[d]thiazol-2(3H)-one 95%

Supplier: Ambeed
Warning

A136145-100MG A136145-1G A136145-250MG A136145-5G
76870-966EA 82.76 USD
76870-966 76870-968 76870-970 76870-972
5-Bromobenzo[d]thiazol-2(3H)-one 95%
5-Bromobenzo[d]thiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrNOS
MW: 230.08 g/mol
MDL Number: MFCD01312116
CAS Number: 199475-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR