CAL-101 98%

Supplier: Ambeed
Warning

Synonyms: Idelalisib, (S)-2-(1-((9H-Purin-6-yl)amino)propyl)-5-fluoro-3-phenylquinazolin-4(3H)-one, GS-1101

A134749-50MG A134749-100MG A134749-10MG A134749-1G A134749-250MG A134749-5G
76870-928EA 17 USD
76870-928 77611-206 77670-934 77670-936 77670-938 77883-088
CAL-101 98%
CAL-101

Precaución: for Research Use Only

Formula: C₂₂H₁₈FN₇O
MDL Number: MFCD19443647
CAS Number: 870281-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR