1-Boc-6-bromoindole-2-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (6-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid

A134746-1G A134746-250MG
76870-924EA 46 USD
76870-924 76870-926
1-Boc-6-bromoindole-2-boronic acid 95%
1-Boc-6-bromoindole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅BBrNO₄
MW: 339.98
MDL Number: MFCD15143453
CAS Number: 1217500-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR