3,4-Bis(difluoromethoxy)benzaldehyde 98%

Supplier: Ambeed
Danger

A133964-10G A133964-1G A133964-25G A133964-5G
76870-878EA 56.23 USD
76870-878 76870-880 76870-882 76870-884
3,4-Bis(difluoromethoxy)benzaldehyde 98%
3,4-Bis(difluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MDL Number: MFCD08706075
CAS Number: 127842-54-0
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR