2-Chloro-6-(1H-pyrazol-1-yl)pyrazine 98%

Supplier: Ambeed
Warning

A133634-1G A133634-25G
76870-872EA 9 USD
76870-872 76870-874
2-Chloro-6-(1H-pyrazol-1-yl)pyrazine 98%
2-Chloro-6-(1H-pyrazol-1-yl)pyrazine

Precaución: for Research Use Only

Formula: C₇H₅ClN₄
MDL Number: MFCD09800958
CAS Number: 642459-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR