5-Bromo-1,3-dimethyl-1H-indazole 98%

Supplier: Ambeed
Warning

A133580-1G A133580-250MG A133580-5G A133580-100MG
76870-868EA 35 USD
76870-868 76870-870 77611-192 77885-148
5-Bromo-1,3-dimethyl-1H-indazole 98%
5-Bromo-1,3-dimethyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MDL Number: MFCD12028623
CAS Number: 552331-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR