3-Chloro-1-(4-nitrophenyl)-5,6-dihydropyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A133226-10G A133226-1G A133226-25G A133226-5G
76870-846EA 63.66 USD
76870-846 76870-848 76870-850 76870-852
3-Chloro-1-(4-nitrophenyl)-5,6-dihydropyridin-2(1H)-one 95%
3-Chloro-1-(4-nitrophenyl)-5,6-dihydropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₉ClN₂O₃
MW: 252.65 g/mol
MDL Number: MFCD18207128
CAS Number: 536760-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR