(R)-1-(3,4-Difluorophenyl)ethanamine 95%

Supplier: Ambeed
Warning

A133117-100MG A133117-1G A133117-250MG
76870-840EA 18 USD
76870-840 76870-842 76870-844
(R)-1-(3,4-Difluorophenyl)ethanamine 95%
(R)-1-(3,4-Difluorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₉F₂N·ClH
MDL Number: MFCD06761760
CAS Number: 321318-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR