Methyl 3-aminocyclohexanecarboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A133105-100MG
A133105-250MG
76870-836EA
12.74
USD
76870-836
76870-838
Methyl 3-aminocyclohexanecarboxylate hydrochloride 95%
Methyl 3-aminocyclohexanecarboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₆ClNO₂ MW: 193.67 g/mol |
MDL Number:
MFCD22392158 CAS Number: 712317-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...