5-Bromo-1,3-thiazole-2-carbaldehyde 98%

Supplier: Ambeed
Warning

A133020-250MG A133020-1G A133020-100MG
76870-830EA 55.17 USD
76870-830 77549-598 77670-892
5-Bromo-1,3-thiazole-2-carbaldehyde 98%
5-Bromo-1,3-thiazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₄H₂BrNOS
MW: 192.03 g/mol
MDL Number: MFCD09870032
CAS Number: 933752-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR