2-(Chloromethyl)benzoic acid 98%

Supplier: Ambeed
Danger

A132766-1G A132766-250MG A132766-25G
76870-818EA 25 USD
76870-818 76870-820 77883-070
2-(Chloromethyl)benzoic acid 98%
2-(Chloromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD06208434
CAS Number: 85888-81-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR