Elvitegravir 98%

Supplier: Ambeed
Warning

Synonyms: EVG, JTK-303/GS-9137, (S)-6-(3-Chloro-2-fluorobenzyl)-1-(1-hydroxy-3-methylbutan-2-yl)-7-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 6-[(3-chloro-2-fluorophenyl)methyl]-1-[(2S)-1-hydroxy-3-methylbutan-2-yl]-7-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

A131621-100MG A131621-1MG A131621-50MG A131621-5MG A131621-250MG A131621-1G
76870-760EA 90.18 USD
76870-760 76870-762 76870-764 76870-766 77592-516 77670-858
Elvitegravir 98%
Elvitegravir

Precaución: for Research Use Only

Formula: C₂₃H₂₃ClFNO₅
MW: 447.88 g/mol
MDL Number: MFCD11846135
CAS Number: 697761-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR