N-(2-Chloroethyl)propan-2-amine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A131310-100MG
A131310-250MG
A131310-5G
76870-758EA
28.65
USD
76870-758
77670-854
77670-856
N-(2-Chloroethyl)propan-2-amine hydrochloride 95%
N-(2-Chloroethyl)propan-2-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClN·ClH MW: 158.069 g/mol |
MDL Number:
MFCD00034904 CAS Number: 6306-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...