4-(4-Ethylpiperazin-1-ylmethyl)benzylamine 97%

Supplier: Ambeed
Warning

A131157-250MG A131157-5G A131157-1G
76870-754EA 42 USD
76870-754 76870-756 77670-852
4-(4-Ethylpiperazin-1-ylmethyl)benzylamine 97%
4-(4-Ethylpiperazin-1-ylmethyl)benzylamine

Precaución: for Research Use Only

Formula: C₁₄H₂₃N₃
MDL Number: MFCD06797801
CAS Number: 914349-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR