4-(Trifluoromethoxy)phenol 98%
Supplier: Ambeed
|
Danger
|
A131146-100G
A131146-1G
A131146-25G
76870-740EA
35
USD
76870-740
76870-742
76870-744
76870-746
77549-562
4-(Trifluoromethoxy)phenol 98%
4-(Trifluoromethoxy)phenol
Precaución: for Research Use Only
Formula:
C₇H₅F₃O₂ |
MDL Number:
MFCD00040988 CAS Number: 828-27-3 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...