2-Chloro-4,4'-bipyridine 95%

Supplier: Ambeed
Warning

A130810-100MG A130810-1G A130810-250MG
76870-716EA 38 USD
76870-716 76870-718 76870-720
2-Chloro-4,4'-bipyridine 95%
2-Chloro-4,4'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂
MW: 190.629 g/mol
MDL Number: MFCD11111098
CAS Number: 53344-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR