6-Chloroimidazo[1,2-b]pyridazine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A130510-100MG A130510-250MG
76870-698EA 27.59 USD
76870-698 76870-700
6-Chloroimidazo[1,2-b]pyridazine-3-carboxylic acid 95%
6-Chloroimidazo[1,2-b]pyridazine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MDL Number: MFCD11870499
CAS Number: 1208084-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR