2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester 97%

Supplier: Ambeed
Warning

A130106-100MG A130106-250MG A130106-1G
76870-678EA 49 USD
76870-678 76870-680 77883-045
2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester 97%
2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD18633018
CAS Number: 152533-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR