2-(Piperidin-1-ylsulfonyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

A128799-100MG A128799-1G A128799-250MG
76870-632EA 15 USD
76870-632 76870-634 76870-636
2-(Piperidin-1-ylsulfonyl)phenylboronic acid 97%
2-(Piperidin-1-ylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄S
MDL Number: MFCD09475844
CAS Number: 957034-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR