1-tert-Butylpyrazole-4-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-t-Butylpyrazole-4-boronic acid, pinacol ester, 1-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A128505-250MG A128505-5G A128505-1G
76870-628EA 16 USD
76870-628 77549-532 77611-126
1-tert-Butylpyrazole-4-boronic acid, pinacol ester 97%
1-tert-Butylpyrazole-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₃BN₂O₂
MDL Number: MFCD16660302
CAS Number: 1256359-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR