Ethyl 2-((methoxycarbonyl)amino)acetate 95%

Supplier: Ambeed
Warning

A128262-1G A128262-5G
76870-622EA 25 USD
76870-622 76870-624
Ethyl 2-((methoxycarbonyl)amino)acetate 95%
Ethyl 2-((methoxycarbonyl)amino)acetate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
MDL Number: MFCD03384670
CAS Number: 5602-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR