5-Fluoro-2-nitropyridin-3-ol 95%

Supplier: Ambeed
Warning

A127783-250MG A127783-1G
76870-610EA 210.06 USD
76870-610 77611-108
5-Fluoro-2-nitropyridin-3-ol 95%
5-Fluoro-2-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃FN₂O₃
MW: 158.09 g/mol
MDL Number: MFCD11878539
CAS Number: 847902-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR