(3-Methylisoxazol-5-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A127577-100MG A127577-250MG
76870-602EA 113.52 USD
76870-602 76870-604
(3-Methylisoxazol-5-yl)methanamine hydrochloride 97%
(3-Methylisoxazol-5-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂O
MW: 148.59 g/mol
MDL Number: MFCD11506173
CAS Number: 70183-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR