Methyl-2,4-dibromobenzoate 97%

Supplier: Ambeed
Warning

A127370-5G A127370-1G A127370-10G
76870-580EA 104 USD
76870-580 77670-800 77883-029
Methyl-2,4-dibromobenzoate 97%
Methyl-2,4-dibromobenzoate

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MDL Number: MFCD09954350
CAS Number: 54335-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR