(S)-2-(4-((((S)-2-Amino-5-methyl-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, calcium salt 98%

Supplier: Ambeed
Warning

Synonyms: Levomefolate calcium

A126968-10MG A126968-1G A126968-1MG A126968-250MG A126968-50MG A126968-5G A126968-25G
76870-558EA 7 USD
76870-558 76870-560 76870-562 76870-564 76870-566 76870-568 77611-094
(S)-2-(4-((((S)-2-Amino-5-methyl-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, calcium salt 98%
(S)-2-(4-((((S)-2-Amino-5-methyl-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, calcium salt

Precaución: for Research Use Only

Formula: C₂₀H₂₃CaN₇O₆
MW: 497.51 g/mol
MDL Number: MFCD15146358
CAS Number: 151533-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR