2-Hydroxyethyl methacrylate 97% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A126933-1000G A126933-100G A126933-500G A126933-25G
76870-552EA 27 USD
76870-552 76870-554 76870-556 77313-118
2-Hydroxyethyl methacrylate 97% stabilized
2-Hydroxyethyl methacrylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MDL Number: MFCD00002863
CAS Number: 868-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR