2-(Pyridin-3-yloxy)ethanamine 97%

Supplier: Ambeed
Warning

A126719-100MG A126719-1G A126719-250MG A126719-5G
76870-536EA 30.77 USD
76870-536 76870-538 76870-540 76870-542
2-(Pyridin-3-yloxy)ethanamine 97%
2-(Pyridin-3-yloxy)ethanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.1671 g/mol
MDL Number: MFCD12192564
CAS Number: 310880-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR