5-Acetyl-2-fluorobenzonitrile 96%

Supplier: Ambeed
Warning

A126310-1G A126310-250MG
76870-514EA 36 USD
76870-514 76870-516
5-Acetyl-2-fluorobenzonitrile 96%
5-Acetyl-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₆FNO
MDL Number: MFCD09831931
CAS Number: 288309-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR