2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester

A126235-100MG A126235-1G A126235-250MG A126235-5G
76870-506EA 13.8 USD
76870-506 76870-508 76870-510 76870-512
2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BFO₄
MW: 294.13 g/mol
MDL Number: MFCD12026092
CAS Number: 944317-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR