2,4-Dichloro-3-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A126130-100MG A126130-1G A126130-5G
76870-498EA 40.32 USD
76870-498 76870-500 76870-502
2,4-Dichloro-3-hydroxybenzaldehyde 95%
2,4-Dichloro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MW: 191.011 g/mol
MDL Number: MFCD22199563
CAS Number: 56962-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR