(2S,3R)-Benzyl 2-(((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Thr-Obzl

A126005-10G A126005-1G A126005-25G A126005-5G
76870-474EA 74.27 USD
76870-474 76870-476 76870-478 76870-480
(2S,3R)-Benzyl 2-(((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate 95%
(2S,3R)-Benzyl 2-(((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅
MW: 431.48 g/mol
MDL Number: MFCD11505901
CAS Number: 73724-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR