2-Bromo-3,4,5-trichloropyridine 95%

Supplier: Ambeed
Warning

A125631-1G A125631-250MG
76870-470EA 33.95 USD
76870-470 76870-472
2-Bromo-3,4,5-trichloropyridine 95%
2-Bromo-3,4,5-trichloropyridine

Precaución: for Research Use Only

Formula: C₅HBrCl₃N
MW: 261.33 g/mol
MDL Number: MFCD22493416
CAS Number: 1330061-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR